Learn More
Chemglass Life Sciences Glass Dish and Plate, 2.8 L Buffer Tray for Capillary Gel Blotting
SDP
Catalog No. 502733282
Encompass_Preferred
Glass Dish and Plate, 2.8 L is a glass buffer tray designed for capillary gel blotting applications such as Northern and Southern blotting. The tray supports wicking paper and a blotting weight, and provides a chemically resistant surface for capillary transfers.
  • Provides 2.8 L buffer capacity (approx. 2800 mL).
  • Constructed from glass for chemical resistance and easy cleaning.
  • Supports wicking paper and blotting weight for capillary transfers.
  • Dimensions: 225 mm x 330 mm x 50 mm.
  • Sold individually (each).
Catalog No. 50-273-3282 Supplier Chemglass Life Sciences Supplier No. CLS1997028
Glass Dish and Plate, 2.8 L is a glass buffer tray designed for capillary gel blotting applications such as Northern and Southern blotting. The tray supports wicking paper and a blotting weight, and provides a chemically resistant surface for capillary transfers.
  • Provides 2.8 L buffer capacity (approx. 2800 mL).
  • Constructed from glass for chemical resistance and easy cleaning.
  • Supports wicking paper and blotting weight for capillary transfers.
  • Dimensions: 225 mm x 330 mm x 50 mm.
  • Sold individually (each).