Learn More
Metrohm USA Separate pH glass electrode
SDP
Catalog No. NC1498328 Shop All Metrohm USA Products
Encompass
pH glass electrode for aqueous pH measurements or differential potentiometry in non-aqueous media. This pH electrode has to be used in combination with a reference electrode.- Optimized length for sample changer applications. - Electrically shielded - Blue T-glass for reliable results Storage in deion. water.
Catalog No. NC1498328 Supplier Metrohm USA Supplier No. 60150100
pH glass electrode for aqueous pH measurements or differential potentiometry in non-aqueous media. This pH electrode has to be used in combination with a reference electrode.- Optimized length for sample changer applications. - Electrically shielded - Blue T-glass for reliable results Storage in deion. water.