Learn More
Vee Gee Vee Gee Glass Erlenmeyer Flask, 300 mL
Catalog No. NC1937424
Encompass_Preferred
The Erlenmeyer Flask features heavy-duty rim to reduce chipping, white permanent graduations, and marking squares. It is made from high-quality borosilicate tubing with a low coefficient of expansion. Uniform wall thickness - ideal for heating applications Graduation accuracy is 5% of full capacity Rubber Stopper No.: 6 Graduation Range: 100 to 300 mL Graduation Intervals: 25 mL
Catalog No. NC1937424 Supplier Vee Gee Supplier No. 10530300A
The Erlenmeyer Flask features heavy-duty rim to reduce chipping, white permanent graduations, and marking squares. It is made from high-quality borosilicate tubing with a low coefficient of expansion. Uniform wall thickness - ideal for heating applications Graduation accuracy is 5% of full capacity Rubber Stopper No.: 6 Graduation Range: 100 to 300 mL Graduation Intervals: 25 mL