Learn More
Alkali Scientific Glass ESR Pipette, Graduated, 300mm, 200/Case
SDP
Supplier:  Alkali Scientific ESRPG
Encompass_Preferred
This 300mm Glass ESR Pipette is designed for accurate and reliable erythrocyte sedimentation rate (ESR) testing in clinical laboratories. Manufactured from high quality, transparent glass, it provides excellent clarity for reading sedimentation levels and ensuring dependable, reproducible results. The pipette features bold, easy to read graduations up to 195mm with 1mm divisions, allowing precise measurement throughout the entire test period. Reusable when properly cleaned and dried, this glass ESR pipette offers a durable and cost effective solution for labs performing routine ESR determinations. Its rigid construction maintains dimensional stability, ensuring consistent performance across repeated uses. Key Features: 300mm length with graduations up to 195mm 1mm graduation divisions for accurate measurement Made from high clarity, durable glass Suitable for standard Westergren ESR testing Reusable when washed and dried correctly Provides precise, reliable sedimentation readings.
Catalog No. 50-006-62055