Learn More
Hach Company Glass Fiber Pads for Moisture Analysis, 200/bx
Supplier:  Hach Company 2507600
Encompass
Used in cases where samples are liquid-like in nature. Glass fiber pads disperse the liquid for even heat distribution. 200/box. 89mm. (Pan PN 2948600)
Catalog No. 50-199-6114