Learn More
Velp™ Scientific Glass Fiber Thimbles for Solvent Extractor
SDP
Catalog No. 11392137
Change view
Click to view available options
Diameter (Metric) Inside:
25 mm
33 mm
40 mm
For Use With (Equipment):
SER148, SER158 Solvent Extractors
SER158/3 Solvent Extractor
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inside For Use With (Equipment)
11-392-137 40 mm SER158/3 Solvent Extractor
11-392-100 25 mm SER158/3 Solvent Extractor
11-392-099 33 mm SER148, SER158 Solvent Extractors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 11-392-137 Supplier Velp™ Scientific Supplier No. A00000393
Add to Cart
Add to Cart
This item is not returnable. View return policy
Glass fiber thimbles for use with solvent extractors.
  • Thimbles for use in extraction
Specifications
Quantity 25 Thimbles
Diameter (Metric) Inside 40 mm
Material Glass
For Use With (Equipment) SER158/3 Solvent Extractor
Length (Metric) 80 mm