Learn More
Chemglass Life Sciences 3mm Diameter Pin, Glass Filled PTFE
SDP
Catalog No. CG209650
Encompass_Preferred
3mm Diameter Pin, Glass Filled PTFE. Replacement part for CG2096 series of High Viscosity Agitators. High viscosity agitator, made entirely of PTFE and glass filled PTFE, provides excellent stirring at the bottom of 2L through 20L reactors, especially when heavy slurries are being mixed. Agitator attaches to stir shaft by both a compression fitting and a glass filled PTFE safety pin. Heavy duty blades are 7mm thick. Supplied complete with one glass filled PTFE safety pin.
Catalog No. CG209650 Supplier Chemglass Life Sciences Supplier No. CG209650
3mm Diameter Pin, Glass Filled PTFE. Replacement part for CG2096 series of High Viscosity Agitators. High viscosity agitator, made entirely of PTFE and glass filled PTFE, provides excellent stirring at the bottom of 2L through 20L reactors, especially when heavy slurries are being mixed. Agitator attaches to stir shaft by both a compression fitting and a glass filled PTFE safety pin. Heavy duty blades are 7mm thick. Supplied complete with one glass filled PTFE safety pin.