Learn More
Advantec MFS, Inc GLASS FILTER GA55 110MM 100/PK
Supplier:  Advantec MFS, Inc GA55110MM
Encompass_Preferred
Glass Fiber Filters made of pure borosilicate glass. Resistant to chemical attack and biologically inert. Use for water/air pollution analysis, clarification or prefilter. No Binder; Nominal Retention 0.6um; Thickness 0.21mm (other grades)
Catalog No. NC0286092