Learn More
Biodesign Inc of Ny Filtration Unit for 47mm circles, Type A, One unit, borosicate glass and stainless steel support
Catalog No. NC2684798
Encompass
Filtration unit for 47mm CellMicroSieve filters. Entire unit with the CellMicroSieve filter in it can be auoclaved.
Catalog No. NC2684798 Supplier Biodesign Inc of Ny Supplier No. F103
Filtration unit for 47mm CellMicroSieve filters. Entire unit with the CellMicroSieve filter in it can be auoclaved.