Learn More
Mettler Toledo™ Glass-free Draft Shield
Catalog No. 01919118 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Glass-free Draft Shield
Catalog No.
01-919-118 Glass-free Draft Shield
1 options
Catalog No. 01-919-118 Supplier Mettler Toledo™ Supplier No. 30409092
Add to Cart
Add to Cart
For XPR/XSR balances with 1 mg/0.1 mg readability
  • Glass-free Draft Shield Pro
  • Fast Stabilization Times
  • For XPR/XSR Balances with 1 mg/0.1 mg readability
  • Patented draft shield doors do not slide open beyond the balance
  • Protects weighing samples from surrounding influences
  • Minimizes air turbulence
  • Ensures faster stabilization
  • Food Industry Compliant - HACCP Standards
  • Saves valuable space within laboratories
  • Height: 248 mm
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) XPR/XSR Balances with 1 mg/0.1 mg Readability
For Use With (Application) To Protect Weighing Samples From Surrounding Influences
Type Glass-Free Draft Shield
Autoclavable Not Autoclavable