Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glass Funnel with Vent
SCI_ED

Catalog No. S34579
Change view
Click to view available options
Diameter (Metric)Top:
9 cm
11.5 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)Top
S34579 9 cm
S34580 11.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S34579 Supplier Variety Glass Supplier No. 6954OZ

Heavy molded-glass funnel with vent.

  • Give displaced air a way to escape from the receiving container so liquid can flow in smoothly
  • Made with smooth rims and inside rib
  • The air vent on the stem makes pouring easy

Specifications

Diameter (Metric)Top 9 cm
Capacity (Metric) 125 mL
Material Glass
For Use With (Application) Gives displaced air a way to escape from the receiving container so liquid can flow in smoothly
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.