Learn More
MSC Mersen Miniature Fuse, Fast-Acting, 32VAC, 20A, Glass Body, 5 ea
Catalog No. 501920188
Encompass
The Mersen GGC20 is a miniature, fast-acting fuse designed to protect sensitive equipment from overcurrents. This glass-body fuse features a 20A ampere rating and a 32V voltage rating, making it suitable for various low-voltage AC applications.
  • Glass body construction
  • Fast-acting characteristic for sensitive equipment protection
  • Rated 20 A at 32 VAC
  • Measures 1/4 in. x 1 1/4 in.
  • UL Listed and CSA Certified
  • Non-rejecting clip mount type
Catalog No. 50-192-0188 Supplier MSC Supplier No. 54008842
The Mersen GGC20 is a miniature, fast-acting fuse designed to protect sensitive equipment from overcurrents. This glass-body fuse features a 20A ampere rating and a 32V voltage rating, making it suitable for various low-voltage AC applications.
  • Glass body construction
  • Fast-acting characteristic for sensitive equipment protection
  • Rated 20 A at 32 VAC
  • Measures 1/4 in. x 1 1/4 in.
  • UL Listed and CSA Certified
  • Non-rejecting clip mount type