Learn More
World Precision Instrument Glass Handling Forceps
SDP
Catalog No. 502521223
Encompass
Ever had difficulty picking up a glass capillary? Special tips on these forceps solve the problem, holding glass firmly without risk of breakage. They also keep the glass clean and avoid contamination from skin oils.
Catalog No. 50-252-1223 Supplier World Precision Instrument Supplier No. 77020
Ever had difficulty picking up a glass capillary? Special tips on these forceps solve the problem, holding glass firmly without risk of breakage. They also keep the glass clean and avoid contamination from skin oils.