Learn More
Bellco Glass, Inc. Glass Impeller Shaft,100-250mL
SDP
Catalog No. 501945163
Encompass_Preferred
Glass mc impeller shaft 7mm od x 155mm long Type 1 Class A boro for 100-250ml spinners. MADE IN THE USAGlass ImpellerShaft,100-250mL
Catalog No. 50-194-5163 Supplier Bellco Glass, Inc. Supplier No. 196970250
Glass mc impeller shaft 7mm od x 155mm long Type 1 Class A boro for 100-250ml spinners. MADE IN THE USAGlass ImpellerShaft,100-250mL