Learn More
Bellco Glass, Inc. Glass Impeller Shaft,15-36L Microcarrier
SDP
Catalog No. 501945075
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. For spinner flasks 15l to 36l. MADE IN THE USA Dimensions: 9mm x 500mm long
Catalog No. 50-194-5075 Supplier Bellco Glass, Inc. Supplier No. 196571500
Manufactured from type 1 class a Borosilicate glass. For spinner flasks 15l to 36l. MADE IN THE USA Dimensions: 9mm x 500mm long