Learn More
Bellco Glass, Inc. Glass Impeller Shaft, 1L
SDP
Catalog No. 501945165
Encompass_Preferred
Type 1 Class A Borosilicate glass 7mm od x 230mm long for mc impellerassemblies. MADE IN THE USAGlass Impeller Shaft,1L
Catalog No. 50-194-5165 Supplier Bellco Glass, Inc. Supplier No. 196971000
Type 1 Class A Borosilicate glass 7mm od x 230mm long for mc impellerassemblies. MADE IN THE USAGlass Impeller Shaft,1L