Learn More
Bellco Glass, Inc. Glass Impeller Shaft,25-100mL
SDP
Catalog No. 501945161
Encompass_Preferred
Description: Glass Impeller Shaft. MADE IN THE USASize: 25-100mLFor Bellco 25-100mL Spinner Flasks & Impeller AssembliesMaterial: Type 1 Class A Borosilicate GlassDimensions: 7mm OD x 120mm Long
Catalog No. 50-194-5161 Supplier Bellco Glass, Inc. Supplier No. 196970100
Description: Glass Impeller Shaft. MADE IN THE USASize: 25-100mLFor Bellco 25-100mL Spinner Flasks & Impeller AssembliesMaterial: Type 1 Class A Borosilicate GlassDimensions: 7mm OD x 120mm Long