Learn More
Bellco Glass, Inc. Glass Impeller Shaft, 3L Micro-Carrier
SDP
Catalog No. 501945166
Encompass_Preferred
Type 1 Class A Borosilicate glass 7mm od x 300mm long Type 1 Class A boro for3L spinners. MADE IN THE USAGlass Impeller Shaft,3L Micro-Carrier
Catalog No. 50-194-5166 Supplier Bellco Glass, Inc. Supplier No. 196973000
Type 1 Class A Borosilicate glass 7mm od x 300mm long Type 1 Class A boro for3L spinners. MADE IN THE USAGlass Impeller Shaft,3L Micro-Carrier