Learn More
Bellco Glass, Inc. Glass Insert,1.0mL
SDP
Catalog No. 501945620
Encompass_Preferred
Inserts are preassembled with a polymer bottom spring. The polymer spring acts as a shock absorber that protects against breakage if the needle bottoms out. The conical design permits complete sample removal. Conical bottom Size1.0ml
Catalog No. 50-194-5620 Supplier Bellco Glass, Inc. Supplier No. 451001010
Inserts are preassembled with a polymer bottom spring. The polymer spring acts as a shock absorber that protects against breakage if the needle bottoms out. The conical design permits complete sample removal. Conical bottom Size1.0ml