Learn More
Microsolv Technology Corporation Inserts, Low Volume, Glass. Clear, 1000ul with a wide, conical point for use in the Micro-Sample Management System. Replacement Inserts. 100/PK.
Catalog No. 502270557
Encompass_Preferred
Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass made for use with the U-2D Micro-Sample Management System Rack and have outer dimensions of 7.72mm x 44.85mm. Also available pre-loaded in U-2D Brand racks of 96.
This item is not returnable. View return policy
Catalog No. 50-227-0557 Supplier Microsolv Technology Corporation Supplier No. 95296I1ML
Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass made for use with the U-2D Micro-Sample Management System Rack and have outer dimensions of 7.72mm x 44.85mm. Also available pre-loaded in U-2D Brand racks of 96.