Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, 100ul fill volume with a conical, precision point and 5x31mm outer dimensions. For use in 8mm, narrow mouth, 2ml autosampler vials. 100/PK.
Supplier:  Microsolv Technology Corporation 9503S05N
Encompass_Preferred
First hydrolytic, Class A, Type 33, Borosilicate Glass. Features a conical tip for excellent performance. Residual volume is 3ul with most autosamplers with a 50ul useable volume. This self aligning insert (no spring required) is slightly taller than other inserts and centers itself with the bottom of the insert, sitting on the bottom of the vial. This product is manufactured in an ISO 9001:2015 Certified facility. Use with 8mm, narrow mouth, Screw Top and 11mm Serum Finish, Crimp Top 2ml, 12x32mm autosampler vials. Economical conical point for budget minded laboratories.
Catalog No. 50-227-0543