Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, 250ul fill volume with a conical, precision point and 6x38mm outer dimensions. For use in 4ml autosampler vials. 100/PK.
Catalog No. 502270544
Encompass_Preferred
First hydrolytic, Class A, Type 33, Borosilicate Glass. Features a mandrel manufactured, precision point conical tip for maximum recovery of sample and internal consistency from insert to insert. Residual volume is 3ul with most autosamplers with a 150ul useable volume. This product is manufactured in an ISO 9001:2015 Certified facility. Springs may be required and are sold separately. Use with 13mm Screw Top 15x45mm autosampler vials.
This item is not returnable. View return policy
Catalog No. 50-227-0544 Supplier Microsolv Technology Corporation Supplier No. 9504S01N
First hydrolytic, Class A, Type 33, Borosilicate Glass. Features a mandrel manufactured, precision point conical tip for maximum recovery of sample and internal consistency from insert to insert. Residual volume is 3ul with most autosamplers with a 150ul useable volume. This product is manufactured in an ISO 9001:2015 Certified facility. Springs may be required and are sold separately. Use with 13mm Screw Top 15x45mm autosampler vials.