Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, Deactivated, 300ul fill volume with a conical point and 6x38mm outer dimensions with plastic spring attached. For use in 4ml autosampler vials. 100/PK.
Catalog No. 502270552
Encompass_Preferred
First hydrolytic, Class A, Type 33, Deactivated / Silanized Borosilicate Glass. Features a conical tip for excellent performance. Residual volume is 3ul with most autosamplers with a 200ul useable volume. Polypropylene springs are attached to keep the inserts center aligned and reduce chances of the needle breaking the inserts during use. This product is manufactured in an ISO 9001:2015 Certified facility. Use with 4ml, 13mm, Screw Top, 15x45mm autosampler vials.
This item is not returnable. View return policy
Catalog No. 50-227-0552 Supplier Microsolv Technology Corporation Supplier No. 9504S04ND
First hydrolytic, Class A, Type 33, Deactivated / Silanized Borosilicate Glass. Features a conical tip for excellent performance. Residual volume is 3ul with most autosamplers with a 200ul useable volume. Polypropylene springs are attached to keep the inserts center aligned and reduce chances of the needle breaking the inserts during use. This product is manufactured in an ISO 9001:2015 Certified facility. Use with 4ml, 13mm, Screw Top, 15x45mm autosampler vials.