Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a flat bottom and 5x31mm outer dimensions. For use in 2ml autosampler vials. 100/PK.
Catalog No. 502271343
Encompass_Preferred
First hydrolytic, Class A, Type N51A, Borosilicate Glass. Features a flat bottom. Residual volume is 8ul with most autosamplers. Use with standard opening, 8mm Screw Top or 11mm Serum Finish Crimp Top 2ml, 12x32mm autosampler vials. Lower cost for budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-1343 Supplier Microsolv Technology Corporation Supplier No. 9582S05NK
First hydrolytic, Class A, Type N51A, Borosilicate Glass. Features a flat bottom. Residual volume is 8ul with most autosamplers. Use with standard opening, 8mm Screw Top or 11mm Serum Finish Crimp Top 2ml, 12x32mm autosampler vials. Lower cost for budget minded laboratories.