Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a flat bottom and 6x31mm outer dimensions. For use in 2ml autosampler vials. . 1000/CS.
Catalog No. 502271344
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass. Features a flat bottom. Residual volume is 6ul with most autosamplers. Use with 9mm Screw Top or 11mm Snap Top 2ml, 12x32mm autosampler vials. Lowest cost for budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-1344 Supplier Microsolv Technology Corporation Supplier No. 9503S05FB
First hydrolytic, Class A, Borosilicate Glass. Features a flat bottom. Residual volume is 6ul with most autosamplers. Use with 9mm Screw Top or 11mm Snap Top 2ml, 12x32mm autosampler vials. Lowest cost for budget minded laboratories.