Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, Deactivated, 350ul fill volume with a flat bottom and 6x31mm outer dimensions. For use in 2ml autosampler vials. 100/PK.
Catalog No. 502270554
Encompass_Preferred
First hydrolytic, Class A, Type 33, Deactivated / Silanized Borosilicate Glass. Features a flat bottom. Residual volume is 6ul with most autosamplers. This product is manufactured in an ISO 9001:2015 Certified facility. Use with 9mm Screw Top or 11mm Snap Top 2ml, 12x32mm autosampler vials. Lower cost for budget minded laboratories.
Catalog No. 50-227-0554 Supplier Microsolv Technology Corporation Supplier No. 9502S05ND
First hydrolytic, Class A, Type 33, Deactivated / Silanized Borosilicate Glass. Features a flat bottom. Residual volume is 6ul with most autosamplers. This product is manufactured in an ISO 9001:2015 Certified facility. Use with 9mm Screw Top or 11mm Snap Top 2ml, 12x32mm autosampler vials. Lower cost for budget minded laboratories.