Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, Deactivated, 350ul fill volume with a flat bottom and 6x31mm outer dimensions. For use in 2ml autosampler vials. 1000/CS.
Catalog No. 502270555
Encompass_Preferred
First hydrolytic, Class A, Type 33, Deactivated / Silanized Borosilicate Glass. Features a flat bottom. Residual volume is 6ul with most autosamplers. This product is manufactured in an ISO 9001:2015 Certified facility. Use with 9mm Screw Top or 11mm Snap Top 2ml, 12x32mm autosampler vials. Lower cost for budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-0555 Supplier Microsolv Technology Corporation Supplier No. 9502S05NDK
First hydrolytic, Class A, Type 33, Deactivated / Silanized Borosilicate Glass. Features a flat bottom. Residual volume is 6ul with most autosamplers. This product is manufactured in an ISO 9001:2015 Certified facility. Use with 9mm Screw Top or 11mm Snap Top 2ml, 12x32mm autosampler vials. Lower cost for budget minded laboratories.