Learn More
Microsolv Technology Corporation Inserts, Low Volume, Glass. Clear, 350ul with a wide, conical point and 6x29mm outer dimensions for use in the Micro-Sample Management System. Replacement Inserts. 100/PK.
Catalog No. 502270558
Encompass_Preferred
Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass made for use with the U-2D Micro-Sample Management System Rack or with 12x32mm, 2ml AQ Brand autosampler vials. Wide conical points help with fast filling and minimizes bubble formation in the insert.
This item is not returnable. View return policy
Catalog No. 50-227-0558 Supplier Microsolv Technology Corporation Supplier No. 9529605N
Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass made for use with the U-2D Micro-Sample Management System Rack or with 12x32mm, 2ml AQ Brand autosampler vials. Wide conical points help with fast filling and minimizes bubble formation in the insert.