Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, 350ul fill volume with a conical point and 6x31mm outer dimensions. For use in 2ml autosampler vials. 1000/CS.
Catalog No. 502271345
Encompass_Preferred
First hydrolytic, Class A, Type N51A, Borosilicate Glass. Features a conical tip for excellent performance. Residual volume is 4ul with most autosamplers. Spring may be required, sold separately, Can be used in 9mm Screw Top or 11mm Snap Top, 2ml 12x32mm autosampler vials. Lowest cost in class for budget minded labs.
This item is not returnable. View return policy
Catalog No. 50-227-1345 Supplier Microsolv Technology Corporation Supplier No. 9502S02LK
First hydrolytic, Class A, Type N51A, Borosilicate Glass. Features a conical tip for excellent performance. Residual volume is 4ul with most autosamplers. Spring may be required, sold separately, Can be used in 9mm Screw Top or 11mm Snap Top, 2ml 12x32mm autosampler vials. Lowest cost in class for budget minded labs.