Learn More
Microsolv Technology Corporation Inserts, Low Volume, Glass. Clear, 500ul with a wide, conical point for use in the Micro-Sample Management System. Replacement Inserts. 100/PK.
Catalog No. 502270559
Encompass_Preferred
Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass made for use with the U-2D Micro-Sample Management System Rack and have outer dimensions of 7.72mm x 31.30mmAlso available pre-loaded in U-2D Brand Racks of 96.
This item is not returnable. View return policy
Catalog No. 50-227-0559 Supplier Microsolv Technology Corporation Supplier No. 95296I500
Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass made for use with the U-2D Micro-Sample Management System Rack and have outer dimensions of 7.72mm x 31.30mmAlso available pre-loaded in U-2D Brand Racks of 96.