Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, 700ul fill volume with a conical, precision point and 8x40mm outer dimensions with plastic spring attached. For use in 4ml autosampler vials. 100/PK.
Catalog No. 502270549
Encompass_Preferred
First hydrolytic, Class A, Type 33, Borosilicate Glass. Features a precision point conical tip for excellent recovery of sample. Residual volume is 8ul with most autosamplers with a 500ul useable volume. Polypropylene springs are attached to keep the inserts center aligned and reduce chances of the needle breaking the inserts during use. This product is manufactured in an ISO 9001:2015 Certified facility. Use with 4ml, 13mm, Screw Top, 15x45mm autosampler vials.
This item is not returnable. View return policy
Catalog No. 50-227-0549 Supplier Microsolv Technology Corporation Supplier No. 9504S03N
First hydrolytic, Class A, Type 33, Borosilicate Glass. Features a precision point conical tip for excellent recovery of sample. Residual volume is 8ul with most autosamplers with a 500ul useable volume. Polypropylene springs are attached to keep the inserts center aligned and reduce chances of the needle breaking the inserts during use. This product is manufactured in an ISO 9001:2015 Certified facility. Use with 4ml, 13mm, Screw Top, 15x45mm autosampler vials.