Learn More
Electron Microscopy Sciences EMS Oscillating Tissue Slicer Glass Knife Holder
SDP
Catalog No. 5025750
Encompass_Preferred
Glass Knife Holder for EMS 4500 and 5000 Oscillating tisue slicer Allows you to reproducibly cut fresh (10 um sections) and fixed tissue, this holder attaches directly to the unit and will accommodate 6.4mm x25mm glass knives.
Catalog No. 50-257-50 Supplier Electron Microscopy Sciences Supplier No. 93038
Glass Knife Holder for EMS 4500 and 5000 Oscillating tisue slicer Allows you to reproducibly cut fresh (10 um sections) and fixed tissue, this holder attaches directly to the unit and will accommodate 6.4mm x25mm glass knives.