Learn More
Electron Microscopy Sciences Glass Knife Pliers
SDP
Catalog No. 5026126
Encompass_Preferred
These pliers are specially designed with its jaws ground to a special shape for ease of preparation when making glass knives.
Catalog No. 50-261-26 Supplier Electron Microscopy Sciences Supplier No. 6219001
These pliers are specially designed with its jaws ground to a special shape for ease of preparation when making glass knives.