Learn More
Electron Microscopy Sciences Glass Knife Scoring Jig Set
SDP
Catalog No. 5026117
Encompass_Preferred
These set of jigs assist in the production of making glass knives by hand. T-square jig: when the edge of this jig is placed against the edge of the glass knife strip the subsequent score will be 26 mm or 1" long. After breaking, the square of the glass can bescored at a 45 degree angle by using the other jig.
Catalog No. 50-261-17 Supplier Electron Microscopy Sciences Supplier No. 6219202
These set of jigs assist in the production of making glass knives by hand. T-square jig: when the edge of this jig is placed against the edge of the glass knife strip the subsequent score will be 26 mm or 1" long. After breaking, the square of the glass can bescored at a 45 degree angle by using the other jig.