Learn More
Ted Pella Inc GLASS KNIFE STATION FOR 8MM
SDP
Catalog No. NC2259645
Encompass
Glass Knife Station, for 8mm thickness. Can be stored dust-free in this container having a silicone rubber mat with conveniently shaped recesses for the knives. Recesses are numbered. Holds 16 knives.
Catalog No. NC2259645 Supplier Ted Pella Inc Supplier No. 8093
Add to Cart
Add to Cart
Glass Knife Station, for 8mm thickness. Can be stored dust-free in this container having a silicone rubber mat with conveniently shaped recesses for the knives. Recesses are numbered. Holds 16 knives.