Learn More
Electron Microscopy Sciences Tripod Polisher Glass Leveling Slide
SDP
Catalog No. 50980882
Encompass
The Glass Leveling Slide (#50116-70): is used for leveling the 3 micrometer's feet or 2 of the feet and the sample. - For use with Tripod Polisher Model 590.
Catalog No. 50-980-882 Supplier Electron Microscopy Sciences Supplier No. 5011670
The Glass Leveling Slide (#50116-70): is used for leveling the 3 micrometer's feet or 2 of the feet and the sample. - For use with Tripod Polisher Model 590.