Learn More
Bellco Glass, Inc. Glass Liquid Additon Assembly ,18mm For 18mm Port
SDP
Catalog No. 501946457
Encompass_Preferred
Glass liquid addition assembly for use on 45mm side arms. Assembly simply screws on and seals with a silicone liner. Small glass chamber for adding liquids to a vessel without-back flow contamination.--One glass addition tube Type 1 Class A Borosilicate. MADE IN THE USA--One compression cap--One front and rear nylon ferrule--One compression fitting--One blue pp screw cap and white silicone liner
Catalog No. 50-194-6457 Supplier Bellco Glass, Inc. Supplier No. 790900060
Glass liquid addition assembly for use on 45mm side arms. Assembly simply screws on and seals with a silicone liner. Small glass chamber for adding liquids to a vessel without-back flow contamination.--One glass addition tube Type 1 Class A Borosilicate. MADE IN THE USA--One compression cap--One front and rear nylon ferrule--One compression fitting--One blue pp screw cap and white silicone liner