Learn More
Bellco Glass, Inc. Glass LV Insert W/Spring,300覘 Bottom Spring
SDP
Catalog No. 501945406
Encompass_Preferred
Inserts are preassembled with a polymer bottom spring. The polymer spring acts as a shock absorbed that protects against breakage if the needle bottoms out. The conical design permits complete sample removal. MADE IN THE USA. Conical bottom Sizes: 300
Catalog No. 50-194-5406 Supplier Bellco Glass, Inc. Supplier No. 207004200
Inserts are preassembled with a polymer bottom spring. The polymer spring acts as a shock absorbed that protects against breakage if the needle bottoms out. The conical design permits complete sample removal. MADE IN THE USA. Conical bottom Sizes: 300