Learn More
Bellco Glass, Inc. Glass LV Insert W/Spring 50
SDP
Supplier:  Bellco Glass, Inc. 207004600
Encompass_Preferred
Inserts are preassembled with a polymer bottom spring. The polymer spring acts as a shock absorbed that protects against breakage if the needle bottoms out. The conical design permits complete sample removal. MADE IN THE USA. Flat bottom Sizes: 50ul
Catalog No. 50-194-5410