Learn More
MilliporeSigma™ Supelco™ Glass Magnet™ Mat
Catalog No. 111009803 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
11-100-9803 1 Ea.
1 options
Catalog No. 11-100-9803 Supplier MilliporeSigma™ Supelco™ Supplier No. 57269
Add to Cart
Add to Cart
Supelco™'s Glass Magnet Sheet prevents your glass apparatus from being knocked over accidentally.
Supelco™'s Glass Magnet Sheet prevents your glass apparatus from being knocked over accidentally. This 2 x 2 ft. sheet (61 cm x 61 cm), with its tacky surface, effectively secures the vessels in place. It can be cut with a razor blade or utility knife to fit strategically on counter tops or in drawers containing fragile glassware. Add it to carts when transporting glass to another lab or into the field.
TRUSTED_SUSTAINABILITY
Specifications
Description W L thickness 24 in. 24 in. 3/16 in.
Quantity 1 Ea.
Glass Magnet is a trademark of Sigma-Aldrich Co. LLC