Learn More
Ace Glass, Inc. Glass manifold assembly with PTFE adapters and 316 stainless steel pressure valve adjustable from 3 to 50 PSI. Intended for small jacketed glassware applications requiring overpressure protection. can be used with gas or liquid.