Learn More
Foxx Life Sciences PUREGRIP Reusable Round Glass Media Storage Bottles with GL45 VersaCap, Pouring Ring, and Extra Large Marking Spot, 250mL, 10/CS
Catalog No. 501542803
Encompass_Preferred
Foxxs PUREGRIP is innovation you can feel. Combining the quality of Borosil glassware with Foxxs user-focused designs, PUREGRIP Bottles offer huge improvements to researchers everyday experience. This new family provides users with the bottles theyve always wanted, without the expensive price tag they might expect. PUREGRIP Media Bottles are easier to open, close, pick up, hold and carry. A frosted surface on the VersaCap allows for labeling with a felt tip marker. A massive enamel surface on the bottle provides even more space. Bottles are resistant to changes in temperature, and the GL45 VersaCap offers the highest rating in the world for a PP cap, 142C. A colorless PP pouring ring allows for drip-free emptying. PUREGRIP Media Bottles are suitable for storing media, buffers, serum, solvents & other materials. All parts are certified USP Class VI. Fully autoclavable. Avail. from 100mL to 20L. The future of glassware is here. Pick it up and see.
Catalog No. 50-154-2803 Supplier Foxx Life Sciences Supplier No. 1501021FLS
Foxxs PUREGRIP is innovation you can feel. Combining the quality of Borosil glassware with Foxxs user-focused designs, PUREGRIP Bottles offer huge improvements to researchers everyday experience. This new family provides users with the bottles theyve always wanted, without the expensive price tag they might expect. PUREGRIP Media Bottles are easier to open, close, pick up, hold and carry. A frosted surface on the VersaCap allows for labeling with a felt tip marker. A massive enamel surface on the bottle provides even more space. Bottles are resistant to changes in temperature, and the GL45 VersaCap offers the highest rating in the world for a PP cap, 142C. A colorless PP pouring ring allows for drip-free emptying. PUREGRIP Media Bottles are suitable for storing media, buffers, serum, solvents & other materials. All parts are certified USP Class VI. Fully autoclavable. Avail. from 100mL to 20L. The future of glassware is here. Pick it up and see.