Learn More
Description
- A drip free pour ring and autoclavable polypropylene (PP) screw cap enable sterilizations of media and reagents.
- Globe Glass™ media bottles are available in sizes 50 mL to 10000 mL.
- ASTM E438, Type I, Class A, Borosilicate 3.3 Glass
- GL32 polypropylene (PP) screw cap on size 50 mL
- GL45 polypropylene (PP) screw caps on sizes 100 mL to 10 L
- Pouring ring for drip free liquid transfer
- High resistance to chemicals
- Dual scale metric graduations
- Durable white enamel printing
- Large white marking area
Specifications
Specifications
| Capacity (Metric) | 1 L |
| Color | Clear |
| Material | Borosilicate Glass (3.3) |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | Yes |
| Mouth | Threaded |
| Closure Size | GL45 |
| Autoclavable | Autoclavable |
| Capacity (English) | 33.81 oz. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.