Learn More
Electron Microscopy Sciences Glass Microbeaker, 1ml, 12 mm diameter, 13 mm height
SDP
Catalog No. 50996334
Encompass_Preferred
For the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
Volume: 1ml
Diameter: 12 mm
Height: 13 mm
Catalog No. 50-996-334 Supplier Electron Microscopy Sciences Supplier No. 60980
Add to Cart
Add to Cart
For the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
Volume: 1ml
Diameter: 12 mm
Height: 13 mm