Learn More
Electron Microscopy Sciences Glass Microbeaker, 5 ml, 25 mm diameter, 19 mm height
SDP
Catalog No. 50996336
Encompass_Preferred
For the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
Volume: 5 ml
Diameter: 25 mm
Height: 19 mm
Catalog No. 50-996-336 Supplier Electron Microscopy Sciences Supplier No. 60984
For the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
Volume: 5 ml
Diameter: 25 mm
Height: 19 mm