Learn More
Electron Microscopy Sciences Glass Microbeaker, 7.5 ml, 25 mm diameter, 25 mm height
SDP
Catalog No. 50996337
Encompass_Preferred
For the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
Volume: 7.5 ml
Diameter: 25 mm
Height: 25 mm
Catalog No. 50-996-337 Supplier Electron Microscopy Sciences Supplier No. 60986
For the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
Volume: 7.5 ml
Diameter: 25 mm
Height: 25 mm