Learn More
World Precision Instrument Glass Microscope Slides, Frosted, Safety Corners, box of 144
SDP
Catalog No. 502539478
Encompass
These clean glass microscope slides are 25 x 75 mm, 1.0~1.2 mm thick with 90° grounded edges. They are available as frosted and red ended. The frosted end slides feature a fine 20 mm frosted area on both sides of one end for easy marking. The red frosted slides feature a 20 mm colored end useful for identifying hazardous materials.
Catalog No. 50-253-9478 Supplier World Precision Instrument Supplier No. 503506
These clean glass microscope slides are 25 x 75 mm, 1.0~1.2 mm thick with 90° grounded edges. They are available as frosted and red ended. The frosted end slides feature a fine 20 mm frosted area on both sides of one end for easy marking. The red frosted slides feature a 20 mm colored end useful for identifying hazardous materials.