Learn More
Bioworld Glass Milk, 5 mL
Catalog No. 50196868
Encompass_Preferred
Glass Milk Specifications:Glass Milk is a specially prepared suspension of propriety silica matrix in an aqueous solution used to isolate and purify DNA. Provides rapid, quantitive, specific binding of DNA/RNA and is able to clean up DNA from solution or clear-up gel bands. DNA purified by Glass Milk shows no effect on sequencing reactions, restriction digests, ligations or phosphorylations. Also suitable for mRNA preparation. Can be stored and shipped at room temperature
Catalog No. 50-196-868 Supplier Bioworld Supplier No. 107600111
Glass Milk Specifications:Glass Milk is a specially prepared suspension of propriety silica matrix in an aqueous solution used to isolate and purify DNA. Provides rapid, quantitive, specific binding of DNA/RNA and is able to clean up DNA from solution or clear-up gel bands. DNA purified by Glass Milk shows no effect on sequencing reactions, restriction digests, ligations or phosphorylations. Also suitable for mRNA preparation. Can be stored and shipped at room temperature