Learn More
Sigma Aldrich Fine Chemicals Biosciences Glass Packed GC Column (fits Agilent, Configuration "A") phase none, matrix 80/100 Porapak Q support, L x O.D. x I.D. 6.0 ft (1.8 m) x 1/4 in. x 2.0 mm |
Catalog No. NC1680804
Encompass
Glass Packed GC Column (fits Agilent, Configuration "A") phase none, matrix 80/100 Porapak Q support, L x O.D. x I.D. 6.0 ft (1.8 m) x 1/4 in. x 2.0 mm |
Catalog No. NC1680804 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 13093U
Glass Packed GC Column (fits Agilent, Configuration "A") phase none, matrix 80/100 Porapak Q support, L x O.D. x I.D. 6.0 ft (1.8 m) x 1/4 in. x 2.0 mm |