Learn More
Sensorex Glass pH Elect,SJ,Refill,BNC
Catalog No. 501897518
Encompass_Preferred
If you deal with a wide variety of chemicals or any aggressive substances, a glass probe like our pH1400 is a safe choice due to the chemically inert properties of glass. This single junction electrode has a refillable reference cell.
Catalog No. 50-189-7518 Supplier Sensorex Supplier No. SG201C
If you deal with a wide variety of chemicals or any aggressive substances, a glass probe like our pH1400 is a safe choice due to the chemically inert properties of glass. This single junction electrode has a refillable reference cell.